1-[3-(Difluoromethoxy)phenyl]-5-oxopyrrolidine-3-carboxylic acid ≥97%

Supplier: Matrix Scientific

082845-1G
102838-242EA 863.08 USD
102838-242
1-[3-(Difluoromethoxy)phenyl]-5-oxopyrrolidine-3-carboxylic acid ≥97%
1-[3-(Difluoromethoxy)phenyl]-5-oxopyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₁F₂NO₄
MDL Number: MFCD20037425
CAS Number: 1246967-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR