1'-Benzyl-5-oxo-1,3'-bipyrrolidine-3-carboxylic acid ≥97%

Supplier: Matrix Scientific

082842-1G
102838-236EA 949.23 USD
102838-236
1'-Benzyl-5-oxo-1,3'-bipyrrolidine-3-carboxylic acid ≥97%
1'-Benzyl-5-oxo-1,3'-bipyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₆H₂₀N₂O₃
MDL Number: MFCD19706012
CAS Number: 1355334-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR