(5-Oxo-1-phenylpyrrolidin-2-yl)acetic acid ≥97%
Supplier: Matrix Scientific
082837-1G
102838-222EA
1287.69
USD
102838-222
(5-Oxo-1-phenylpyrrolidin-2-yl)acetic acid ≥97%
(5-Oxo-1-phenylpyrrolidin-2-yl)acetic acid
Hazard: Irritant
Formula:
C₁₂H₁₃NO₃ |
MDL Number:
MFCD09759164 CAS Number: 88612-68-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...