1-(1-Methylpiperidin-4-yl)pyrrolidine-2-carboxylic acid ≥97%

Supplier: Matrix Scientific

082827-1G
102838-202EA 780 USD
102838-202
1-(1-Methylpiperidin-4-yl)pyrrolidine-2-carboxylic acid ≥97%
1-(1-Methylpiperidin-4-yl)pyrrolidine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₂₀N₂O₂
MDL Number: MFCD11100089
CAS Number: 1401102-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR