1-Cyclohexylpyrrolidine-2-carboxylic acid ≥97%

Supplier: Matrix Scientific

082826-1G
102838-200EA 750.77 USD
102838-200
1-Cyclohexylpyrrolidine-2-carboxylic acid ≥97%
1-Cyclohexylpyrrolidine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₉NO₂
MDL Number: MFCD11100088
CAS Number: 862902-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR