1-(4-Methoxybenzyl)pyrrolidine-3-carboxylic acid ≥97%
Supplier: Matrix Scientific
082820-1G
102838-188EA
806.15
USD
102838-188
1-(4-Methoxybenzyl)pyrrolidine-3-carboxylic acid ≥97%
1-(4-Methoxybenzyl)pyrrolidine-3-carboxylic acid
Hazard: Irritant
Formula:
C₁₃H₁₇NO₃ |
MDL Number:
MFCD09864936 CAS Number: 1086375-34-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...