1-(3-Fluorobenzyl)pyrrolidine-3-carboxylic acid ≥97%
Supplier: Matrix Scientific
082818-1G
102838-184EA
684.62
USD
102838-184
1-(3-Fluorobenzyl)pyrrolidine-3-carboxylic acid ≥97%
1-(3-Fluorobenzyl)pyrrolidine-3-carboxylic acid
Hazard: Irritant
Formula:
C₁₂H₁₄FNO₂ |
MDL Number:
MFCD09864934 CAS Number: 1086375-23-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...