1-(4-Fluorobenzyl)pyrrolidine-3-carboxylic acid ≥97%

Supplier: Matrix Scientific

082817-1G
102838-182EA 806.15 USD
102838-182
1-(4-Fluorobenzyl)pyrrolidine-3-carboxylic acid ≥97%
1-(4-Fluorobenzyl)pyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₄FNO₂
MDL Number: MFCD09864933
CAS Number: 933709-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR