1-(4-Chlorobenzyl)pyrrolidine-3-carboxylic acid ≥97%

Supplier: Matrix Scientific

082815-1G
102838-178EA 806.15 USD
102838-178
1-(4-Chlorobenzyl)pyrrolidine-3-carboxylic acid ≥97%
1-(4-Chlorobenzyl)pyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₄ClNO₂
MDL Number: MFCD09864931
CAS Number: 1086375-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR