1-[4-(Trifluoromethoxy)phenyl]pyrrolidine-3-carboxylic acid ≥97%
Supplier: Matrix Scientific
082814-1G
102838-176EA
806.15
USD
102838-176
1-[4-(Trifluoromethoxy)phenyl]pyrrolidine-3-carboxylic acid ≥97%
1-[4-(Trifluoromethoxy)phenyl]pyrrolidine-3-carboxylic acid
Hazard: Irritant
Formula:
C₁₂H₁₂F₃NO₃ |
MDL Number:
MFCD09864930 CAS Number: 1086375-06-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...