1-(4-Fluorophenyl)pyrrolidine-3-carboxylic acid ≥97%
Supplier: Matrix Scientific
082808-1G
102838-164EA
806.15
USD
102838-164
1-(4-Fluorophenyl)pyrrolidine-3-carboxylic acid ≥97%
1-(4-Fluorophenyl)pyrrolidine-3-carboxylic acid
Hazard: Irritant
Formula:
C₁₁H₁₂FNO₂ |
MDL Number:
MFCD09864924 CAS Number: 933731-71-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...