1-(3-Chlorophenyl)pyrrolidine-3-carboxylic acid ≥97%
Supplier: Matrix Scientific
082807-1G
102838-162EA
806.15
USD
102838-162
1-(3-Chlorophenyl)pyrrolidine-3-carboxylic acid ≥97%
1-(3-Chlorophenyl)pyrrolidine-3-carboxylic acid
Hazard: Irritant
Formula:
C₁₁H₁₂ClNO₂ |
MDL Number:
MFCD09864923 CAS Number: 1086374-92-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...