5-Oxo-1-[4-(trifluoromethoxy)phenyl]pyrrolidine-3-carboxylic acid ≥97%
Supplier: Matrix Scientific
082804-1G
102838-156EA
426.15
USD
102838-156
5-Oxo-1-[4-(trifluoromethoxy)phenyl]pyrrolidine-3-carboxylic acid ≥97%
5-Oxo-1-[4-(trifluoromethoxy)phenyl]pyrrolidine-3-carboxylic acid
Hazard: Irritant
Formula:
C₁₂H₁₀F₃NO₄ MW: 289.21 g/mol |
MDL Number:
MFCD01569312 CAS Number: 874765-55-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...