1-Pyridin-3-ylpyrrolidine-3-carboxylic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(Pyridin-3-yl)pyrrolidine-3-carboxylic acid

082802-1G
102838-152EA 1118.46 USD
102838-152
1-Pyridin-3-ylpyrrolidine-3-carboxylic acid ≥97%
1-Pyridin-3-ylpyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₂
MW: 192.22 g/mol
MDL Number: MFCD09258862
CAS Number: 1086374-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR