1-[4-(Trifluoromethyl)benzyl]pyrrolidine-3-carboxylic acid ≥97%

Supplier: Matrix Scientific

082796-1G
102838-140EA 806.15 USD
102838-140
1-[4-(Trifluoromethyl)benzyl]pyrrolidine-3-carboxylic acid ≥97%
1-[4-(Trifluoromethyl)benzyl]pyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₄F₃NO₂
MDL Number: MFCD09258857
CAS Number: 1086380-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR