1-Isoxazol-3-yl-5-oxopyrrolidine-3-carboxylic acid ≥97%

Supplier: Matrix Scientific

082792-1G
102838-132EA 949.23 USD
102838-132
1-Isoxazol-3-yl-5-oxopyrrolidine-3-carboxylic acid ≥97%
1-Isoxazol-3-yl-5-oxopyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₈H₈N₂O₄
MDL Number: MFCD08235227
CAS Number: 914637-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR