5-Oxo-1-pyrimidin-2-ylpyrrolidine-3-carboxylic acid ≥97%

Supplier: Matrix Scientific

082789-1G
102838-126EA 750.77 USD
102838-126
5-Oxo-1-pyrimidin-2-ylpyrrolidine-3-carboxylic acid ≥97%
5-Oxo-1-pyrimidin-2-ylpyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₉H₉N₃O₃
MDL Number: MFCD07364773
CAS Number: 944648-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR