1-[6-(Trifluoromethyl)pyridazin-3-yl]piperidine-4-carboxylic acid ≥97%
Supplier: Matrix Scientific
082755-1G
102838-050EA
920
USD
102838-050
1-[6-(Trifluoromethyl)pyridazin-3-yl]piperidine-4-carboxylic acid ≥97%
1-[6-(Trifluoromethyl)pyridazin-3-yl]piperidine-4-carboxylic acid
Hazard: Irritant
Formula:
C₁₁H₁₂F₃N₃O₂ |
MDL Number:
MFCD11840273 CAS Number: 1211534-85-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...