1-[(1-Methyl-1H-imidazol-4-yl)sulfonyl]piperidine-4-carboxylic acid ≥97%

Supplier: Matrix Scientific

082754-1G
102838-048EA 750.77 USD
102838-048
1-[(1-Methyl-1H-imidazol-4-yl)sulfonyl]piperidine-4-carboxylic acid ≥97%
1-[(1-Methyl-1H-imidazol-4-yl)sulfonyl]piperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₅N₃O₄S
MW: 273.31 g/mol
MDL Number: MFCD03408010
CAS Number: 874781-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR