1'-(tert-Butoxycarbonyl)-1,3'-bipiperidine-4-carboxylic acid ≥97%
Supplier: Matrix Scientific
082750-1G
102838-040EA
506.15
USD
102838-040
1'-(tert-Butoxycarbonyl)-1,3'-bipiperidine-4-carboxylic acid ≥97%
1'-(tert-Butoxycarbonyl)-1,3'-bipiperidine-4-carboxylic acid
Hazard: Irritant
Formula:
C₁₆H₂₈N₂O₄ |
MDL Number:
MFCD11100082 CAS Number: 1216759-08-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...