1-(4-Methylpyrimidin-2-yl)piperidine-4-carboxylic acid ≥97%
Supplier: Matrix Scientific
082748-1G
102838-036EA
666.15
USD
102838-036
1-(4-Methylpyrimidin-2-yl)piperidine-4-carboxylic acid ≥97%
1-(4-Methylpyrimidin-2-yl)piperidine-4-carboxylic acid
Hazard: Irritant
Formula:
C₁₁H₁₅N₃O₂ |
MDL Number:
MFCD10703526 CAS Number: 891391-61-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...