1-[6-(Trifluoromethyl)pyridin-2-yl]piperidine-4-carboxylic acid ≥97%

Supplier: Matrix Scientific

082737-1G
102838-010EA 553.85 USD
102838-010
1-[6-(Trifluoromethyl)pyridin-2-yl]piperidine-4-carboxylic acid ≥97%
1-[6-(Trifluoromethyl)pyridin-2-yl]piperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₃F₃N₂O₂
MDL Number: MFCD08235221
CAS Number: 582325-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR