1-(1,3-Thiazol-2-ylmethyl)piperidine-4-carboxylic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
1-(Thiazol-2-ylmethyl)piperidine-4-carboxylic acid
082735-1G
102838-006EA
636.92
USD
102838-006
1-(1,3-Thiazol-2-ylmethyl)piperidine-4-carboxylic acid ≥97%
1-(1,3-Thiazol-2-ylmethyl)piperidine-4-carboxylic acid
Hazard: Irritant
Formula:
C₁₀H₁₄N₂O₂S MW: 226.3 g/mol |
MDL Number:
MFCD08235219 CAS Number: 1086380-54-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...