1-[(2-Methyl-1H-imidazol-4-yl)methyl]piperidine-4-carboxylic acid ≥97%

Supplier: Matrix Scientific

082734-1G
102838-004EA 538.46 USD
102838-004
1-[(2-Methyl-1H-imidazol-4-yl)methyl]piperidine-4-carboxylic acid ≥97%
1-[(2-Methyl-1H-imidazol-4-yl)methyl]piperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₇N₃O₂
MDL Number: MFCD08235218
CAS Number: 1086380-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR