1-(1H-Imidazol-4-ylmethyl)piperidine-4-carboxylic acid ≥97%

Supplier: Matrix Scientific

082733-1G
102838-002EA 636.92 USD
102838-002
1-(1H-Imidazol-4-ylmethyl)piperidine-4-carboxylic acid ≥97%
1-(1H-Imidazol-4-ylmethyl)piperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₅N₃O₂
MDL Number: MFCD08235217
CAS Number: 914637-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR