1-(Thien-3-ylmethyl)piperidine-4-carboxylic acid ≥97%

Supplier: Matrix Scientific

082731-1G
102837-998EA 524.62 USD
102837-998
1-(Thien-3-ylmethyl)piperidine-4-carboxylic acid ≥97%
1-(Thien-3-ylmethyl)piperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₅NO₂S
MDL Number: MFCD05169822
CAS Number: 901923-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR