1-(3-Furylmethyl)piperidine-4-carboxylic acid ≥97%
Supplier: Matrix Scientific
082727-1G
102837-990EA
524.62
USD
102837-990
1-(3-Furylmethyl)piperidine-4-carboxylic acid ≥97%
1-(3-Furylmethyl)piperidine-4-carboxylic acid
Hazard: Irritant
Formula:
C₁₁H₁₅NO₃ |
MDL Number:
MFCD08235215 CAS Number: 914637-42-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...