1-(3-Aminobenzyl)-5-methyl-1H-1,2,3-triazole-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

082723-1G
102837-982EA 750.77 USD
102837-982
1-(3-Aminobenzyl)-5-methyl-1H-1,2,3-triazole-4-carboxylic acid ≥95%
1-(3-Aminobenzyl)-5-methyl-1H-1,2,3-triazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₂N₄O₂
MDL Number: MFCD09864919
CAS Number: 1086380-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR