7-(Trifluoromethyl)-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid ≥97%

Supplier: Matrix Scientific

082718-1G
102837-970EA 1118.46 USD
102837-970
7-(Trifluoromethyl)-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid ≥97%
7-(Trifluoromethyl)-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₈F₃NO₃
MDL Number: MFCD09258855
CAS Number: 1086380-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR