6-Hydroxy-2-(methoxycarbonyl)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid ≥97%
Supplier: Matrix Scientific
082715-1G
102837-962EA
636.92
USD
102837-962
6-Hydroxy-2-(methoxycarbonyl)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid ≥97%
6-Hydroxy-2-(methoxycarbonyl)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
Hazard: Irritant
Formula:
C₁₂H₁₃NO₅ MW: 251.24 g/mol |
MDL Number:
MFCD11505020 CAS Number: 1219199-11-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...