[(5,6-Dichloro-1-ethyl-1H-benzimidazol-2-yl)thio]acetic acid ≥97%

Supplier: Matrix Scientific

082694-1G
102837-904EA 1118.46 USD
102837-904
[(5,6-Dichloro-1-ethyl-1H-benzimidazol-2-yl)thio]acetic acid ≥97%
[(5,6-Dichloro-1-ethyl-1H-benzimidazol-2-yl)thio]acetic acid

Hazard: Irritant

Formula: C₁₁H₁₀Cl₂N₂O₂S
MDL Number: MFCD00106534
CAS Number: 263148-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR