3-Hydroxy-1-benzothiophene-2-carboxylic acid ≥97%

082679-1G
102837-864EA 645.15 USD
102837-864
3-Hydroxy-1-benzothiophene-2-carboxylic acid ≥97%
3-Hydroxy-1-benzothiophene-2-carboxylic acid

Hazard: Irritant

Formula: C₉H₆O₃S
MDL Number: MFCD00460995
CAS Number: 191031-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR