5-(3-Bromophenyl)-1,2,4-oxadiazole-3-carboxylic acid ≥97%
Supplier: Matrix Scientific
082676-1G
102837-858EA
806.15
USD
102837-858
5-(3-Bromophenyl)-1,2,4-oxadiazole-3-carboxylic acid ≥97%
5-(3-Bromophenyl)-1,2,4-oxadiazole-3-carboxylic acid
Hazard: Irritant
Formula:
C₉H₅BrN₂O₃ |
MDL Number:
MFCD09027678 CAS Number: 1216977-92-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...