3-(4-Bromobenzyl)-1,2,4-oxadiazole-5-carboxylic acid ≥97%

Supplier: Matrix Scientific

082673-1G
102837-852EA 636.92 USD
102837-852
3-(4-Bromobenzyl)-1,2,4-oxadiazole-5-carboxylic acid ≥97%
3-(4-Bromobenzyl)-1,2,4-oxadiazole-5-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₇BrN₂O₃
MDL Number: MFCD07379488
CAS Number: 1215585-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR