3-(2-Bromobenzyl)-1,2,4-oxadiazole-5-carboxylic acid ≥97%

Supplier: Matrix Scientific

082672-1G
102837-850EA 636.92 USD
102837-850
3-(2-Bromobenzyl)-1,2,4-oxadiazole-5-carboxylic acid ≥97%
3-(2-Bromobenzyl)-1,2,4-oxadiazole-5-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₇BrN₂O₃
MDL Number: MFCD07379490
CAS Number: 1216634-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR