5-(3-Furyl)-1,2,4-oxadiazole-3-carboxylic acid ≥97%

Supplier: Matrix Scientific

082669-1G
102837-844EA 949.23 USD
102837-844
5-(3-Furyl)-1,2,4-oxadiazole-3-carboxylic acid ≥97%
5-(3-Furyl)-1,2,4-oxadiazole-3-carboxylic acid

Hazard: Irritant

Formula: C₇H₄N₂O₄
MDL Number: MFCD09258854
CAS Number: 1086380-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR