(2,6-Dimethyltetrahydro-2H-pyran-4-yl)acetic acid ≥97%

Supplier: Matrix Scientific

082661-1G
102837-830EA 920 USD
102837-830
(2,6-Dimethyltetrahydro-2H-pyran-4-yl)acetic acid ≥97%
(2,6-Dimethyltetrahydro-2H-pyran-4-yl)acetic acid

Hazard: Irritant

Formula: C₉H₁₆O₃
MDL Number: MFCD11100081
CAS Number: 1215417-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR