3-(Pyrrolidin-1-yl)pyrazine-2-carboxylic acid ≥97%

Supplier: Matrix Scientific

082647-1G
102837-794EA 949.23 USD
102837-794
3-(Pyrrolidin-1-yl)pyrazine-2-carboxylic acid ≥97%
3-(Pyrrolidin-1-yl)pyrazine-2-carboxylic acid

Hazard: Irritant

Formula: C₉H₁₁N₃O₂
MDL Number: MFCD08056310
CAS Number: 342422-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR