1-Methyl-2-propyl-1H-imidazole-4-carboxylic acid ≥97%

Supplier: Matrix Scientific

082642-1G
102837-784EA 1118.46 USD
102837-784
1-Methyl-2-propyl-1H-imidazole-4-carboxylic acid ≥97%
1-Methyl-2-propyl-1H-imidazole-4-carboxylic acid

Hazard: Irritant

Formula: C₈H₁₂N₂O₂
MDL Number: MFCD19706022
CAS Number: 1355334-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR