1-Methyl-2-propyl-1H-imidazole-4,5-dicarboxylic acid ≥97%

Supplier: Matrix Scientific

082641-1G
102837-782EA 949.23 USD
102837-782
1-Methyl-2-propyl-1H-imidazole-4,5-dicarboxylic acid ≥97%
1-Methyl-2-propyl-1H-imidazole-4,5-dicarboxylic acid

Hazard: Irritant

Formula: C₉H₁₂N₂O₄
MDL Number: MFCD19706020
CAS Number: 881422-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR