3-Methyl-5-(4-methyl-1,2,3-thiadiazol-5-yl)-1-phenyl-1H-pyrazole-4-carboxylic acid ≥97%

Supplier: Matrix Scientific

082631-1G
102837-762EA 1118.46 USD
102837-762
3-Methyl-5-(4-methyl-1,2,3-thiadiazol-5-yl)-1-phenyl-1H-pyrazole-4-carboxylic acid ≥97%
3-Methyl-5-(4-methyl-1,2,3-thiadiazol-5-yl)-1-phenyl-1H-pyrazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₂N₄O₂S
MDL Number: MFCD00662733
CAS Number: 264880-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR