3-tert-Butyl-1H-pyrazole-4-carboxylic acid ≥97%
Supplier: Matrix Scientific
082624-1G
102837-746EA
566.15
USD
102837-746
3-tert-Butyl-1H-pyrazole-4-carboxylic acid ≥97%
3-tert-Butyl-1H-pyrazole-4-carboxylic acid
Hazard: Irritant
Formula:
C₈H₁₂N₂O₂ MW: 168.2 g/mol |
MDL Number:
MFCD09258851 CAS Number: 1152540-42-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...