2-Methyl-4-phenyl-1,3-oxazole-5-carboxylic acid ≥97%

Supplier: Matrix Scientific

082618-1G
102837-734EA 920 USD
102837-734
2-Methyl-4-phenyl-1,3-oxazole-5-carboxylic acid ≥97%
2-Methyl-4-phenyl-1,3-oxazole-5-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₉NO₃
Melting Pt: 221…223 °C
MDL Number: MFCD01562926
CAS Number: 51143-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR