3-tert-Butylisoxazole-4-carboxylic acid ≥97%
Supplier: Matrix Scientific
082600-1G
102837-694EA
750.77
USD
102837-694
3-tert-Butylisoxazole-4-carboxylic acid ≥97%
3-tert-Butylisoxazole-4-carboxylic acid
Hazard: Irritant
Formula:
C₈H₁₁NO₃ |
MDL Number:
MFCD11100075 CAS Number: 1217047-14-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...