5-(2-Furyl)-1,3-thiazole-4-carboxylic acid ≥97%
Supplier: Matrix Scientific
082579-1G
102837-648EA
636.92
USD
102837-648
5-(2-Furyl)-1,3-thiazole-4-carboxylic acid ≥97%
5-(2-Furyl)-1,3-thiazole-4-carboxylic acid
Hazard: Irritant
Formula:
C₈H₅NO₃S |
MDL Number:
MFCD09991776 CAS Number: 1086380-33-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...