2-(2-Bromobenzyl)-1,3-thiazole-4-carboxylic acid ≥97%
Supplier: Matrix Scientific
082566-1G
102837-622EA
553.85
USD
102837-622
2-(2-Bromobenzyl)-1,3-thiazole-4-carboxylic acid ≥97%
2-(2-Bromobenzyl)-1,3-thiazole-4-carboxylic acid
Hazard: Irritant
Formula:
C₁₁H₈BrNO₂S |
MDL Number:
MFCD09258847 CAS Number: 1086380-14-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...