5-(5-Formylpyridin-2-yl)thiophene-2-carboxylic acid ≥97%

Supplier: Matrix Scientific

082523-1G
102837-522EA 750.77 USD
102837-522
5-(5-Formylpyridin-2-yl)thiophene-2-carboxylic acid ≥97%
5-(5-Formylpyridin-2-yl)thiophene-2-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₇NO₃S
MDL Number: MFCD09800585
CAS Number: 844495-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR