5-(6-Formylpyridin-2-yl)thiophene-2-carboxylic acid ≥97%

082522-1G
102837-520EA 561.2 USD
102837-520
5-(6-Formylpyridin-2-yl)thiophene-2-carboxylic acid ≥97%
5-(6-Formylpyridin-2-yl)thiophene-2-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₇NO₃S
MDL Number: MFCD09800584
CAS Number: 656227-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR