5-Pyrimidin-5-ylthiophene-2-carboxylic acid ≥97%

Supplier: Matrix Scientific

082518-1G
102837-512EA 949.23 USD
102837-512
5-Pyrimidin-5-ylthiophene-2-carboxylic acid ≥97%
5-Pyrimidin-5-ylthiophene-2-carboxylic acid

Hazard: Irritant

Formula: C₉H₆N₂O₂S
MDL Number: MFCD06802393
CAS Number: 893738-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR