5-(1-Methylimidazol-2-yl)thiophene-2-carboxylic acid ≥97%

Supplier: Matrix Scientific

082509-1G
102837-492EA 949.23 USD
102837-492
5-(1-Methylimidazol-2-yl)thiophene-2-carboxylic acid ≥97%
5-(1-Methylimidazol-2-yl)thiophene-2-carboxylic acid

Hazard: Irritant

Formula: C₉H₈N₂O₂S
MDL Number: MFCD08056306
CAS Number: 914637-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR